Anti-AGPAT3 Rabbit Polyclonal Antibody [clone: 432CT31.4.2]

Supplier: Abgent
ABGEAP8686CEA 407 CHF
ABGEAP8686C
Anti-AGPAT3 Rabbit Polyclonal Antibody [clone: 432CT31.4.2]
Antibodies
Type: Primary
Antigen: AGPAT3 (1-acylglycerol-3-phosphate O-acyltransferase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR