Anti-KIR3DS1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8673BEA 405 CHF
ABGEAP8673B
Anti-KIR3DS1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KIR3DS1 (killer cell immunoglobulin-like receptor, three domains, short cytoplasmic tail, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR