Anti-ITGB3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8672BEA 378 CHF
ABGEAP8672B
Anti-ITGB3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ITGB3 (integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR