Anti-FLCN Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8658BEA 405 CHF
ABGEAP8658B
Anti-FLCN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FLCN (folliculin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR