Anti-BECN1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8653AEA 448 CHF
ABGEAP8653A
Anti-BECN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: BH3 domain
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR