Anti-SLC23A2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8652AEA 407 CHF
ABGEAP8652A
Anti-SLC23A2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC23A2 (solute carrier family 23 (ascorbic acid transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR