Anti-CAV2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8645AEA 405 CHF
ABGEAP8645A
Anti-CAV2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAV2 (caveolin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR