Anti-EHHADH Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8636BEA 405 CHF
ABGEAP8636B
Anti-EHHADH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EHHADH (enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR