Anti-KLRC1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8629BEA 405 CHF
ABGEAP8629B
Anti-KLRC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLRC1 (killer cell lectin-like receptor subfamily C, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR