Anti-MICA Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8626CEA 431 CHF
ABGEAP8626C
Anti-MICA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MICA (MHC class I polypeptide-related sequence A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR