Anti-RNF5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8608AEA 405 CHF
ABGEAP8608A
Anti-RNF5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RNF5 (ring finger protein 5, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR