Anti-VTI1A Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8602BEA 407 CHF
ABGEAP8602B
Anti-VTI1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: VTI1A (vesicle transport through interaction with t-SNAREs 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR