Anti-PPP2R2B Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8600CEA 407 CHF
ABGEAP8600C
Anti-PPP2R2B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP2R2B (protein phosphatase 2, regulatory subunit B, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR