Anti-SERPINB7 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8597CEA 405 CHF
ABGEAP8597C
Anti-SERPINB7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SERPINB7 (serpin peptidase inhibitor, clade B (ovalbumin), member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR