Anti-SRC Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8596AEA 407 CHF
ABGEAP8596A
Anti-SRC Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SRC (v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR