Anti-USF2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8585CEA 405 CHF
ABGEAP8585C
Anti-USF2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: USF2 (upstream transcription factor 2, c-fos interacting)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR