Anti-CCL3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8527BEA 419 CHF
ABGEAP8527B
Anti-CCL3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCL3 (chemokine (C-C motif) ligand 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR