Anti-NR1H2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8526AEA 378 CHF
ABGEAP8526A
Anti-NR1H2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NR1H2 (nuclear receptor subfamily 1, group H, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR