Anti-ARHGAP18 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8516CEA 448 CHF
ABGEAP8516C
Anti-ARHGAP18 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ARHGAP18 (Rho GTPase activating protein 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR