Anti-PLA2G4A Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8510CEA 419 CHF
ABGEAP8510C
Anti-PLA2G4A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PLA2G4A (phospholipase A2, group IVA (cytosolic, calcium-dependent))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR