Anti-GPD1 Rabbit Polyclonal Antibody [clone: 445CT13.3.3]

Supplier: Abgent
ABGEAP8507AEA 407 CHF
ABGEAP8507A
Anti-GPD1 Rabbit Polyclonal Antibody [clone: 445CT13.3.3]
Antibodies
Type: Primary
Antigen: GPD1 (glycerol-3-phosphate dehydrogenase 1 (soluble))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR