Anti-TPT1 Rabbit Polyclonal Antibody [clone: 362CT19.3.3]
Supplier: Abgent
ABGEAP8505CEA 419 CHF
ABGEAP8505C
Anti-TPT1 Rabbit Polyclonal Antibody [clone: 362CT19.3.3]
Antibodies
Type: Primary
Antigen: TPT1 (tumor protein, translationally-controlled 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human