Anti-PPP2R1A Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8462AEA 448 CHF
ABGEAP8462A
Anti-PPP2R1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP2R1A (protein phosphatase 2, regulatory subunit A, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR