Anti-DUSP14 Rabbit Polyclonal Antibody [clone: 428CT12.2.6.4]

Lieferant: Abgent
ABGEAP8456AEA 405 CHF
ABGEAP8456A
Anti-DUSP14 Rabbit Polyclonal Antibody [clone: 428CT12.2.6.4]
Antikörper
Type: Primary
Antigen: DUSP14 (dual specificity phosphatase 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR