Anti-DUSP13-L184 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8455BEA 378 CHF
ABGEAP8455B
Anti-DUSP13-L184 Rabbit Polyclonal Antibody
Type: Primary
Antigen: DUSP13-L184
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR