Anti-DUSP4 Rabbit Polyclonal Antibody [clone: 358CT10.3.6]

Supplier: Abgent
ABGEAP8447BEA 407 CHF
ABGEAP8447B
Anti-DUSP4 Rabbit Polyclonal Antibody [clone: 358CT10.3.6]
Antibodies
Type: Primary
Antigen: DUSP4 (dual specificity phosphatase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR