Anti-Rat MKP3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8445AEA 405 CHF
ABGEAP8445A
Anti-Rat MKP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Rat MKP3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR