Anti-PLRG1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8439AEA 405 CHF
ABGEAP8439A
Anti-PLRG1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLRG1 (pleiotropic regulator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR