Anti-PPM1D Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8437BEA 419 CHF
ABGEAP8437B
Anti-PPM1D Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPM1D (protein phosphatase, Mg2+/Mn2+ dependent, 1D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR