Anti-PPP1CC Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8432AEA 405 CHF
ABGEAP8432A
Anti-PPP1CC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPP1CC (protein phosphatase 1, catalytic subunit, gamma isozyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR