Anti-PTPN1 Rabbit Polyclonal Antibody [clone: 331CT17.4.4]

Lieferant: Abgent
ABGEAP8411EEA 405 CHF
ABGEAP8411E
Anti-PTPN1 Rabbit Polyclonal Antibody [clone: 331CT17.4.4]
Antikörper
Type: Primary
Antigen: PTPN1 (protein tyrosine phosphatase, non-receptor type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR