Anti-PTPN1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8411CEA 378 CHF
ABGEAP8411C
Anti-PTPN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTPN1 (protein tyrosine phosphatase, non-receptor type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR