Anti-PTPN9 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8408BEA 405 CHF
ABGEAP8408B
Anti-PTPN9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTPN9 (protein tyrosine phosphatase, non-receptor type 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR