Anti-TRPM8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8181BEA 405 CHF
ABGEAP8181B
Anti-TRPM8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRPM8 (transient receptor potential cation channel, subfamily M, member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR