Anti-BAIAP2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8174AEA 448 CHF
ABGEAP8174A
Anti-BAIAP2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BAIAP2 (BAI1-associated protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR