Anti-BAI1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8170AEA 407 CHF
ABGEAP8170A
Anti-BAI1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BAI1 (brain-specific angiogenesis inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR