Anti-BRSK1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8168BEA 448 CHF
ABGEAP8168B
Anti-BRSK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BRSK1 (BR serine/threonine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR