Anti-KIP2 Rabbit Polyclonal Antibody [clone: 366CT28.2.3]

Supplier: Abgent
ABGEAP8158AEA 407 CHF
ABGEAP8158A
Anti-KIP2 Rabbit Polyclonal Antibody [clone: 366CT28.2.3]
Antibodies
Type: Primary
Antigen: KIP2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR