Anti-YWHAZ Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8152CEA 405 CHF
ABGEAP8152C
Anti-YWHAZ Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: YWHAZ (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR