Anti-PFKFB1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8147AEA 378 CHF
ABGEAP8147A
Anti-PFKFB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PFKFB1 (6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR