Anti-CKII alpha (CSNK2A1) Rabbit Polyclonal Antibody
Supplier: Abgent
ABGEAP8144CEA 448 CHF
ABGEAP8144C
Anti-CKII alpha (CSNK2A1) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CKII alpha (CSNK2A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse