Anti-PFKL Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8136BEA 419 CHF
ABGEAP8136B
Anti-PFKL Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PFKL (phosphofructokinase, liver)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR