Anti-PFKP Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8135BEA 419 CHF
ABGEAP8135B
Anti-PFKP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PFKP (phosphofructokinase, platelet)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR