Anti-GSK3A Rabbit Polyclonal Antibody [clone: 396CT20.4.4]

Supplier: Abgent
ABGEAP8120BEA 419 CHF
ABGEAP8120B
Anti-GSK3A Rabbit Polyclonal Antibody [clone: 396CT20.4.4]
Antibodies
Type: Primary
Antigen: GSK3A (glycogen synthase kinase 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR