Anti-ULK3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8115AEA 378 CHF
ABGEAP8115A
Anti-ULK3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ULK3 (unc-51 like kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR