Anti-NPPC Rabbit Polyclonal Antibody [clone: 365CT24.1.1]
Supplier: Abgent
ABGEAP8113AEA 448 CHF
ABGEAP8113A
Anti-NPPC Rabbit Polyclonal Antibody [clone: 365CT24.1.1]
Antibodies
Type: Primary
Antigen: Natriuretic Peptide Receptor C (NPR3/ANPC)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human