Anti-TP53RK Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8097CEA 448 CHF
ABGEAP8097C
Anti-TP53RK Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TP53RK (TP53 regulating kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR