Anti-NEK1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8072BEA 448 CHF
ABGEAP8072B
Anti-NEK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NEK1 (NIMA-related kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR