Anti-TRPM7 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8052BEA 419 CHF
ABGEAP8052B
Anti-TRPM7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRPM7 (transient receptor potential cation channel, subfamily M, member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR