Anti-BRD3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8050BEA 405 CHF
ABGEAP8050B
Anti-BRD3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BRD3 (bromodomain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR